missing translation for 'onlineSavingsMsg'
Learn More
JNJ-10229570, MedChemExpress Product Code.: 16480457

JNJ-10229570, MedChemExpress

Product Code. 16480457
50 mg, 50mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480457

Brand: Medchem Express HY107139

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JNJ-10229570 is an antagonist of melanocortin receptor 1 (MC1R) and melanocortin receptor 5 (MC5R), which inhibits sebaceous gland differentiation and the production of sebum-specific lipids. JNJ-10229570 inhibits the binding of 125I-NDP-α-MSH to cells expressing human MC1R and MC5R, with IC50 values of 270 nM and 200 nM, respectively.

JNJ-10229570 is an antagonist of melanocortin receptor 1 (MC1R) and melanocortin receptor 5 (MC5R), which inhibits sebaceous gland differentiation and the production of sebum-specific lipids. JNJ-10229570 inhibits the binding of 125I-NDP-α-MSH to cells expressing human MC1R and MC5R, with IC50 values of 270 nM and 200 nM, respectively.

Specifications

Chemical Name or Material JNJ-10229570
CAS 524923-88-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H19N3O2S
Quantity 50 mg
Solubility Information DMSO : 62.5 mg/mL (160.47 mM; Need ultrasonic)
SMILES COC1=C(C=CC=C1)C(N(C2=C(C=CC=C2)OC)S/3)=NC3=N\C4=CC=CC=C4
Molecular Weight (g/mol) 389.47
Formula Weight 389.47
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
JNJ-10229570, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.