Learn More
Cole-Parmer™ Jenway™ Glass UV Semi Micro Cuvette
Brand: Cole-Parmer™ 83070-86
This item is not returnable.
View return policy
Specifications
UV Semi-micro Cuvette | |
1 mL | |
Quartz | |
10 mm |
Product Content Correction
Product Title
Cole-Parmer™ Jenway™ Glass UV Semi Micro Cuvette > Capacity: 1mL