Learn More
Cole-Parmer™ Jenway™ Glass Cuvettes
1250.00 NOK - 5030.00 NOK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
13197124
|
Cole-Parmer™
99610-30 |
2 to 3.5 mL |
1250.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
13151734
|
Cole-Parmer™
99610-31 |
14 mL |
2060.00 NOK
Each |
Estimated Shipment: 10-12-2025
Log in to see stock. |
||||||
11435019
|
Cole-Parmer™
83070-42 |
35.0 mL |
5030.00 NOK
Each |
Estimated Shipment: 10-12-2025
Log in to see stock. |
||||||
13131734
|
Cole-Parmer™
83070-40 |
7.0 mL |
1810.00 NOK
Each |
Estimated Shipment: 10-12-2025
Log in to see stock. |
||||||
11480569
|
Cole-Parmer™
99610-32 |
17.5 mL |
3180.00 NOK
Each |
Estimated Shipment: 10-12-2025
Log in to see stock. |
||||||
Specifications
Cuvette | |
Glass |
Product Title
Cole-Parmer™ Jenway™ Glass Cuvettes