Learn More
Jenway™ Flame Photometry Standard, 3000ppm Barium
Brand: Jenway 99959-42
Quantity | 500 mL |
---|
This item is not returnable.
View return policy
Specifications
3000 ppm Barium | |
500 mL |
Product Content Correction
Product Title
Jenway™ Flame Photometry Standard, 3000ppm Barium > Quantity: 500mL