Learn More
Jenway™ Conductivity Standards
Click to view available options
Quantity:
500 mL
Conductivity:
12.88 mS
84 μS
Description
- For use with Jenway™ Conductivity and TDS meters.
- Choose 12.88m/S or 84μ/S standards depending on your specific needs
- Available in quantities of 500mL
- For use with Jenway™ Conductivity and TDS meters.
- Choose 12.88m/S or 84μ/S standards depending on your specific needs
- Available in quantities of 500mL
Specifications
Specifications
Conductivity | 84 μS |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Product Title
Jenway™ Conductivity Standards >