Learn More
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
Brand:  GPE Scientific™ CSR/250
 View more versions of this product
Product Code. 15804744
  • 2065.00 NOK / Each
Only null left
Estimated Shipment: 02-09-2025
Description
  • Designed with a side arm for evacuating or filling with gases
  • For connection to Schlenk lines
  • Round-bottomed shape
  • Includes O-ring tap
  • Includes O-ring ball joint cup
Specifications
Specifications
250 mL
O-Ring Tap and O-Ring Ball Joint Cup
Vacuum/Schlenk Flask
Size: 29
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup > Capacity: 250mL