Learn More
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Fine Thread Tap and Socket
Product Code. 15864804
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. 15864804
Brand: GPE Scientific™ JRF/250
Only null left
Add to basket
Add to basket
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Fine Thread Tap and Socket is specially designed for use with Schlenk lines and vacuum manifolds.
  • Solidly constructed for use with vacuum manifolds
  • Designed for use with Schlenk lines
  • Includes fine thread tap and socket
Specifications
Capacity (Metric) 250 mL
For Use With (Equipment) Schlenk lines and vacuum manifolds
Includes Fine thread tap and socket
Socket Joint Size: 24
Bottom Shape Round