Learn More
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap and B14 Socket
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap and B14 Socket is specially designed for use with vacuum manifolds and Schlenk lines.
Brand: GPE Scientific™ JCS/50
This item is not returnable.
View return policy
Description
- Solidly constructed for use with vacuum manifolds
- Designed for use with Schlenk lines
- Includes fine thread tap and B14 socket
Specifications
50 mL | |
Fine thread tap and B14 socket | |
Size: 14 |
Vacuum manifolds and Schlenk lines | |
Sample Flask |
Product Content Correction
Product Title
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap and B14 Socket > Capacity: 50mL