Learn More
Avantor™ J.T. Baker™ Luer PTFE Connectors (one-piece)
Extraction Column Processor Replacement Parts for J.T. Baker™ BAKER spe-12G Column Processor (PTFE design)
Brand: Avantor™ 4586
10393.00 NOK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Spare part for Extraction Column Processor
- Use with J.T. Baker™ BAKER spe-12G Column Processor
Specifications
Connector | |
PTFE | |
12/box |