missing translation for 'onlineSavingsMsg'
Learn More

Ivermectin, ≥95%, MP Biomedicals™

Product Code. 11402651 Shop All MP Biomedicals Products
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ivermectin is a broad-spectrum antiparasitic agent. Ivermectin is a macrolide antiobiotic, & semisynthetic derivative of avermectin B1a and B1b

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70288-86-7
Molecular Formula C48H74O14
Molecular Weight (g/mol) 875.106
MDL Number MFCD00869511
InChI Key AZSNMRSAGSSBNP-RDKIRRGNSA-N
Synonym 22, 23-Dihydroavermectin B1, Hyvermectin
PubChem CID 122172888
SMILES CCC(C)C1C(CCC2(O1)CC3CC(O2)CC=C(C(C(C=CC=C4COC5C4(C(C=C(C5O)C)C(=O)O3)O)C)OC6CC(C(C(O6)C)OC7CC(C(C(O7)C)O)OC)OC)C)C

Specifications

Type Ivermectin
Physical Form Crystalline Powder
Color White or White/Yellow
Quantity 250 mg
Assay Percent Range ≥95%
Formula Weight 875.1
Chemical Name or Material Ivermectin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ivermectin, ≥95%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.