missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITPA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16134-1-AP-150UL
This item is not returnable.
View return policy
Description
Pyrophosphatase that hydrolyzes the non-canonical purine nucleotides inosine triphosphate (ITP), deoxyinosine triphosphate (dITP) as well as 2′-deoxy-N-6-hydroxylaminopurine triposphate (dHAPTP) and xanthosine 5′-triphosphate (XTP) to their respective monophosphate derivatives. The enzyme does not distinguish between the deoxy- and ribose forms. Probably excludes non-canonical purines from RNA and DNA precursor pools, thus preventing their incorporation into RNA and DNA and avoiding chromosomal lesions.Specifications
ITPA | |
Polyclonal | |
Unconjugated | |
ITPA | |
C20orf37, HLC14 06 P, Inosine triphosphatase, ITPA, ITPase, OK/SW cl.9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16434, 311422, 3704 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3ZW55, Q9BY32, Q9D892 | |
Itpa | |
ITPA Fusion Protein Ag9128 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction