missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITLN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11770-1-AP-150UL
This item is not returnable.
View return policy
Description
ITLN1 has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes.ITLN1 increases AKT phosphorylation in the absence and presence of insulin. ITLN1 may play a role in the defense system against microorganisms. ITLN1 may specifically recognize carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues, in a calcium-dependent manner. ITLN1 may be involved in iron metabolism.Specifications
ITLN1 | |
Polyclonal | |
Unconjugated | |
ITLN1 | |
Endothelial lectin HL 1, Galactofuranose binding lectin, hIntL, HL 1, HL1, Intelectin 1, INTL, ITLN, ITLN 1, ITLN1, LFR, omentin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16429, 498284, 55600 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88310, Q8WWA0 | |
Itln1 | |
ITLN1 Fusion Protein Ag2424 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction