Learn More
ITLN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11770-1-AP-20UL
 View more versions of this product
Product Code. 16859063
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
ITLN1 has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes.ITLN1 increases AKT phosphorylation in the absence and presence of insulin. ITLN1 may play a role in the defense system against microorganisms. ITLN1 may specifically recognize carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues, in a calcium-dependent manner. ITLN1 may be involved in iron metabolism.
Specifications
Specifications
ITLN1
Polyclonal
Unconjugated
ITLN1
Endothelial lectin HL 1, Galactofuranose binding lectin, hIntL, HL 1, HL1, Intelectin 1, INTL, ITLN, ITLN 1, ITLN1, LFR, omentin
Rabbit
Antigen Affinity Chromatography
RUO
16429, 498284, 55600
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O88310, Q8WWA0
Itln1
ITLN1 Fusion Protein Ag2424
20 μL
Primary
Mouse, Human, Rat