missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl cinnamate, 98%, Thermo Scientific™

Product Code. 11342067
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7780-06-5
MDL Number MFCD00048292

Specifications

Density 1.032
Boiling Point 268°C to 270°C
Flash Point 94°C (201°F)
Molecular Formula C12H14O2
Refractive Index 1.545
Quantity 100 g
Beilstein 1908938
Synonym isopropyl cinnamate, cinnamic acid, isopropyl ester, iso-propyl cinnamate, 2-propenoic acid, 3-phenyl-, 1-methylethyl ester, 1-methylethyl 3-phenylpropenoate, unii-6rwc10bdd7, isopropyl 3-phenylpropenoate, fema no. 2939, trans-cinnamic acid isopropyl ester, isopropyl 3-phenylacrylate
InChI Key RGACABDFLVLVCT-CMDGGOBGSA-N
SMILES CC(C)OC(=O)C=CC1=CC=CC=C1
IUPAC Name propan-2-yl (E)-3-phenylprop-2-enoate
Molecular Weight (g/mol) 190.24
PubChem CID 5273464
Formula Weight 190.24
Percent Purity 98%
Chemical Name or Material Isopropyl cinnamate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'