missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, 99% (Molecular Biology Reagent), MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Specifications
Specifications
Quantity | 100 mL |
Formula Weight | 60.09 |
Chemical Name or Material | Isopropyl Alcohol, 99% |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Isopropyl Alcohol, 99% (Molecular Biology Reagent), MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction