missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoamyl Propionate, MP Biomedicals
Click to view available options
Quantity:
100 g
Specifications
CAS | 105-68-0 |
Molecular Formula | C8H16O2 |
Quantity | 100 g |
Synonym | isoamyl propionate, isoamyl propanoate, isopentyl propanoate, 3-methylbutyl propionate, isopentyl propionate, isopentyl alcohol, propionate, 1-butanol, 3-methyl-, propanoate, iso-pentyl propionate, propionic acid, isopentyl ester, 3-methyl-1-butyl propanoate |
InChI Key | XAOGXQMKWQFZEM-UHFFFAOYSA-N |
SMILES | CCC(=O)OCCC(C)C |
IUPAC Name | 3-methylbutyl propanoate |
Molecular Weight (g/mol) | 144.214 |
PubChem CID | 7772 |
ChEBI | CHEBI:87419 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'