Learn More
Brady™ ISO Safety Sign without Text - Wash Your Hands
Safety sign for indicating mandatory action.
Brand: Brady™ 819464
44.37 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sign compliant to ISO 7010
- Improves workplace safety
Specifications
PIC M011-Dia 200-PE-CRD/1 | |
Laminated Polyester | |
200 mm | |
White on Blue |
Gloss | |
Permanent Acrylic | |
Round |
Product Content Correction
Product Title
Brady™ ISO Safety Sign without Text - Wash Your Hands > Diameter: 200 mm