Learn More
ISE Buffer Solution Fluoride, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Quantity | 500 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ISEF523F1 | Certificate of Analysis | 29/08/2024 | ISEF5 |
ISEF522M1 | Certificate of Analysis | 03/05/2023 | ISEF5 |
ISEF522B1 | Certificate of Analysis | 23/02/2023 | ISEF5 |
ISEF521H1 | Certificate of Analysis | 24/05/2022 | ISEF5 |
ISEF520C1 | Certificate of Analysis | 23/07/2021 | ISEF5 |
Product Content Correction
Product Title
ISE Buffer Solution Fluoride, Reagecon™ >