Learn More
Hanna Instruments™ Iron Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Iron Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
Brand: Hanna Instruments™ HI93746-01
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determination of iron. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Reagent sets available for low range determination (using the TPTZ method) or high range determination (using USEPA method 315B and Standard Method 3500-Fe B)
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
TPTZ | |
Compatible Hanna benchtop and portable photometers (HI83099, HI83200, HI83205, HI83208, HI96746) | |
100 | |
0 to 1.6mg/L iron | |
Water Testing Kit |
Iron low range determination | |
100 individual packets | |
1 Set | |
Cholorimetric determination |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Reagent kit, 100 packets for 50 tests Iron .
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- EUH031-Contact with acids liberates toxic gas.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : potassium disulphate, Sodium dithionite,2,4,6-Tris(2-pyridyl)-s-triazine