Learn More
BollĂ© Safety IRI-s Platinum™ Clear Safety Glasses Product Code.: 13416979
BollĂ© Safety IRI-s Platinum™ Clear Safety Glasses
Product Code. 13416979
Clear, Each
Click to view available options
Lens Tint:
Clear
Unit Size:
Each
Product Code. 13416979
Brand: Bollé Safety IRIPSI
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured by Bolle Safety the IRI-s Safety Glasses are coated with Platinum™, which is a permanent coating that gives a high scratch resistance and delays fogging beyond the minimum required standards.
  • PLATINUM™ anti-fog and anti-scratch coating
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety glasses
Weight (Metric) 27 g
  • BollĂ© Safety - Platinum coating
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Bolle Safety Platinum coating
    • Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Bolle Safety Platinum efficiency demonstration
      • Video Player is loading.
        Current Time 0:00
        Duration 0:00
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 0:00
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
          Bolle Safety IRI-s
        Product Content Correction
        Product Title
        BollĂ© Safety IRI-s Platinum™ Clear Safety Glasses >