Learn More
Ipatasertib, MedChemExpress
MedChemExpress Ipatasertib (GDC-0068) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively.
Brand:  Medchem Express HY-15186
Encompass_Preferred
 View more versions of this product
Product Code. 16422467
  • 4741.12 NOK / 50mg
Explore more special offers
Description
Description
Ipatasertib (GDC-0068) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively.
Specifications
Specifications
Ipatasertib
1001264-89-6
50 mg
C24H32ClN5O2
GDC-0068 RG7440
ClC1=CC=C([C@@H](CNC(C)C)C(N2CCN(C3=C([C@H](C)C[C@H]4O)C4=NC=N3)CC2)=O)C=C1
458.0
Research
Powder
Off-White
Research
Cancer-Kinase/protease
DMSO : 220 mg/mL (480.35 mM; Need ultrasonic)
458.0
95.0%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ipatasertib, MedChemExpress