missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetic acid, 97%

CAS: 64-69-7 | C2H3IO2 | 185.95 g/mol

966.00 NOK - 2305.00 NOK

Chemical Identifiers

CAS 64-69-7
Molecular Formula C2H3IO2
Molecular Weight (g/mol) 185.95
MDL Number MFCD00002685
InChI Key JDNTWHVOXJZDSN-UHFFFAOYSA-N
Synonym iodoacetic acid, acetic acid, iodo, iodoacetate, monoiodoacetic acid, monoiodoacetate, monoiodine acetate, iodoaceticacid, iodoethanoic acid, acetic acid, 2-iodo, kyselina jodoctova
PubChem CID 5240
ChEBI CHEBI:74571
IUPAC Name 2-iodoacetic acid
SMILES C(C(=O)O)I
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10071260
SDS View Documents
Thermo Scientific Acros
122280250
25 g Glass bottle
966.00 NOK
25g
Please to purchase this item. Need a web account? Register with us today!
10635742
SDS View Documents
Thermo Scientific Acros
122280500
50 g Glass bottle
1535.00 NOK
50g
Please to purchase this item. Need a web account? Register with us today!
10449230
SDS View Documents
Thermo Scientific Acros
122281000
100 g Glass bottle
2305.00 NOK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodoacetic acid, 97%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.