Learn More
Iodine, crystalline, 99.5% Product Code.: 11397817
Iodine, crystalline, 99.5%
Product Code. 11397817
250 g, 250g
Click to view available options
Quantity:
250 g
Unit Size:
250g
Product Code. 11397817
Brand: Thermo Scientific Alfa Aesar 014248.30
Request Bulk or Custom Format
CAS: 7553-56-2 | I2 | 253.81 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
PubChem CID 807
ChEBI CHEBI:17606
SMILES II
Specifications
Melting Point 183°C to 186° (sublimation)
Physical Form Solid
Quantity 250 g
Assay Percent Range 99.5%
MDL Number MFCD00011355 MFCD00164163
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
IUPAC Name molecular iodine
Formula Weight 253.81
Density 4.93
Chemical Name or Material Iodine
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberN27L056 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code014248.A1,014248.30,014248.14
Lot NumberQ02K001 Certificate TypeCertificate of Analysis Date11/06/2024 Product Code014248.A1,014248.30,014248.14
Lot NumberY17I028 Certificate TypeCertificate of Analysis Date24/10/2023 Product Code014248.A1,014248.30,014248.14
Lot NumberW15J042 Certificate TypeCertificate of Analysis Date04/10/2023 Product Code014248.A1,014248.30,014248.14
Lot NumberM26J023 Certificate TypeCertificate of Analysis Date01/02/2023 Product Code014248.A1,014248.30,014248.14
Product Title
Iodine, crystalline, 99.5% >