missing translation for 'onlineSavingsMsg'
Learn More

Iodine 0.02365 M (0.0473 N) Analytical Volumetric Solution (AVL), Reagecon
BIOPHARMA

Product Code. 11526793 Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 11526793

Brand: Reagecon I20023F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fully factorized, high purity, certified, stable product, developed and tested for oxidation/reduction titrations.

  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • Accurate, stable, reproducible and fully factorized.
  • Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
  • Certified to a specification of ± 0.1%.
  • Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
  • May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
  • Oxidation/reduction titrations depend on changes in the electropotential of the oxidation reduction system and always involves a reaction between an oxidizing and reducing pair.

Specifications

Description Accuracy ± 0.01%
Packaging Amber Glass Winchester Bottle GL45
Physical Form Liquid
Concentration 0.02365 M
Technique Titration
Quantity 1 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.