Learn More
Iodate Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™ Product Code.: 12902135
Iodate Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
BIOPHARMA
Product Code. 12902135
100 mL
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Product Code. 12902135
Brand: Fisher Chemical A 021 100
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Quantity 100 mL
Solution Contains H2O
Concentration 1 g/L
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1026466 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberA 021 100
Lot Number1015378 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberA 021 100
Product Content Correction
Product Title
Iodate Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™ >