Learn More
Invitrogen™ pIZT/V5-His Vector Kit
For high-level expression of the gene of interest
Brand: Invitrogen™ V801001
Includes: The InsectSelect Glow Kit includes 20ÎĽg each of supercoiled, lyophilized pIZT/V5-His and pIZT/V5-His/CAT, 2ÎĽg each of lyophilized OpIE2 Forward and Reverse Sequencing primers, 1g Zeocin, Cellfectin Reagent, Sf9 cells, and 1 L of GIBCO Grace's Insect Media. The pIZT/V5-His Vector Kit contains only the expression vectors and primers as described above.
Description
- The OpIE2 promoter for constitutive expression
- The Zeocin resistance gene for rapid selection of stably transfected cell lines
- C-terminal V5 epitope and polyhistidine (6xHis) sequence for detection with Anti-V5 Antibody and rapid purification with nickel-chelating resin
- In addition, pIZT/V5-His expresses a fusion of the green fluorescent protein and the Zeocin resistance protein (Zeo-GFP)
- The Zeo-GFP fusion protein permits rapid selection of stably transfected cell lines with Zeocin and confers a fluorescent phenotype that simplifies identification of transfected cells
Constitutive Protein Expression in Insect Cells, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
OplE2 | |
The InsectSelect™ Glow Kit includes 20 μg each of supercoiled, lyophilized pIZT/V5-His and pIZT/V5-His/CAT, 2 μg each of lyophilized OpIE2 Forward and Reverse Sequencing primers, 1 g Zeocin,™ Cellfectin™ Reagent, Sf9 cells, and 1 L of GIBCO™ Grace's Insect Media. The pIZT/V5-His Vector Kit contains only the expression vectors and primers as described above. Store frozen insect cells in liquid nitrogen. Store medium and Cellfectin™ Reagent at +4°C. Store all other reagents at -20°C. All reagents are guaranteed stable for 6 months when properly stored. | |
His Tag (6x), V5 Epitope Tag | |
1 kit | |
InsectSelect™ |
Insect Cell Expression Vector | |
C-terminal | |
Restriction Enzyme/MCS | |
pIZ |
InsectSelect Glow System with pIZT/V5-His
Version: Version H 10/20/10
File size: 1,008.8 kB
Product Content Correction
Product Title
Invitrogen™ pIZT/V5-His Vector Kit > 1 kit