Learn More
Invitrogen™ pBAD-DEST49 Gateway™ Destination Vector
Designed for rapid cloning with Gateway entry clone using lambda phage site-specific recombination and subsequent high-level, tightly regulated expression in E. coli
Brand:  Invitrogen™ 12283016
Product Code. 11568696
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: The pBAD-DEST49 Gateway destination vector includes 6μg of vector.
Description
Description
  • araBAD promoter for tightly regulated expression in E. coli
  • HP-thioredoxin fusion for improved protein yield and solubility
  • Enterokinase cleavage site for efficient cleavage of N-terminal fusion with EKMax™
  • C-terminal V5 and 6xHis tags for efficient detection and purification of fusion proteins
  • R sites for efficient recombination with any attL-flanked Gateway entry vector
Bacterial Expression/Cloning/Gateway Cloning/Protein Expression/Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Inducible
araBAD
None
Ampicillin (AmpR)
His Tag (6x), V5 Epitope Tag, Thioredoxin
6 μg
Gateway™
Arabinose
Gateway System Destination Expression Vector
The pBAD-DEST49 Gateway™ destination vector includes 6 μg of vector. Store at -20°C. The vector is guaranteed stable for 6 months when properly stored.
EK (Enterokinase) Recognition Site
Gateway
pBAD, pDEST
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Manuals
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Invitrogen™ pBAD-DEST49 Gateway™ Destination Vector > 6μg
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only. Not for use in diagnostic procedures.