Learn More
Invitrogen™ E-Gel™ General Purpose Agarose Gels, 1.2%
Self-contained, precast agarose gels designed to provide fast, convenient, and easy electrophoresis
Brand: Invitrogen™ G501801
Includes: Contains one multi-pack of 18 gels.
Description
- Ready for use with agarose, electrodes, and DNA stain packaged inside a dry, disposable, UV-transparent cassette
- There are no gels to pour, buffers to make, staining or destaining steps to perform, or gel boxes to assemble
- Offer excellent resolution and clarity in 12 minutes and are excellent for analyzing PCR products, restriction digests, and plasmid preparations
- 12-well, single comb gels
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature

Specifications
Contains one multi-pack of 18 gels. Store at room temperature. | |
E-Gel | |
12-well | |
E-Gel™ | |
Single Comb |
1.2% | |
Agarose Gel | |
Ethidium Bromide | |
Room Temperature |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A141221-01 | Certificate of Analysis | 29/01/2025 | G501801 |
A160322-01 | Certificate of Analysis | 29/01/2025 | G501801 |
A020622-01 | Certificate of Analysis | 29/01/2025 | G501801 |
A160622-01 | Certificate of Analysis | 29/01/2025 | G501801 |
A280722-01 | Certificate of Analysis | 29/01/2025 | G501801 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.