Learn More
Invitrogen™ E-Base™ 90°-Angle Adapter
For use with E-Base™ Integrated Devices
Brand: Invitrogen™ EA03
1567.70 NOK valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Description
E-Base™ devices run E-Gel™ and E-PAGE™ buffer-less, pre-cast agarose and polyacrylamide gels. This adapter allows the use of E-Gel and E-PAGE gels on automated liquid handling workstations. The adapter is positioned on the automated workstation and allows the gripper to turn the gels by 90 degrees to accommodate different pipette attachments, configurations, and technologies.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
E-PAGE™, E-Base™ | |
E-Base Angle Adapter | |
1 adapter |
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.