Learn More
Invitrogen™ E-Base™ 90°-Angle Adapter
For use with E-Base™ Integrated Devices
Brand:  Invitrogen™ EA03
1567.70 NOK valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 15381527
  • 2570.00 NOK / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
E-Base™ devices run E-Gel™ and E-PAGE™ buffer-less, pre-cast agarose and polyacrylamide gels. This adapter allows the use of E-Gel and E-PAGE gels on automated liquid handling workstations. The adapter is positioned on the automated workstation and allows the gripper to turn the gels by 90 degrees to accommodate different pipette attachments, configurations, and technologies.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E-PAGE™, E-Base™
E-Base Angle Adapter
1 adapter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.