Learn More
Cytiva INV-907 Injection Kit
Range of manual and automated valves to meet variety of performance requirements.
Brand: Cytiva 18-1110-89
This item is not returnable.
View return policy
Description
- Designed for biocompatibility
- Each valve may be inserted into flow path without causing unnecessary mixing or sample dilution
- Includes fill port and needle
Specifications
Injection Kit | |
ETFE, Titanium | |
Kit includes fill port and needle. |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Product Content Correction
Product Title
Cytiva INV-907 Injection Kit >