Learn More
Cole-Parmer™ Integrity 10, RS-200, and RS-250 Reaction Station Accessories
Keep your Integrity 10, RS-200, or RS-250 Reaction Station running smoothly with these parts and accessories.
4300.00 NOK - 76795.00 NOK
Specifications
For Use With (Equipment) Integrity 10
View More Specs
Products 6
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11718342
View Documents
Cole-Parmer™
36630-85
Glass Tubes, 24 mm x 150 mm
4300.00 NOK
Pack of 10
Only null left
Estimated Shipment: 14-07-2025
Add to basket
11798332
View Documents
Cole-Parmer™
36630-92
Integrity Software
62110.00 NOK
Each
Only null left
Estimated Shipment: 14-07-2025
Add to basket
12719048
View Documents
Cole-Parmer™
36630-88
Small Intrusive IR probe PID-NIR5-BNSD
76795.00 NOK
Each
Only null left
Estimated Shipment: 14-07-2025
Add to basket
11896121
View accessory products View Documents
Cole-Parmer™
36630-80
Multi Temp Module
42075.00 NOK
Each
Only null left
Estimated Shipment: 14-07-2025
Add to basket
12621116
View Documents
Cole-Parmer™
36630-89
Multi IR Box
57100.00 NOK
Each
Only null left
Estimated Shipment: 01-07-2025
Add to basket
12709048
View Documents
Cole-Parmer™
36630-90
Non Intrusive IR Sensor
29020.00 NOK
Each
Only null left
Estimated Shipment: 14-07-2025
Add to basket
Description
Description
  • Suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations.
  • Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software.
Specifications
Specifications
Integrity 10
Documents
Product Certifications
Product Content Correction
Product Title
Cole-Parmer™ Integrity 10, RS-200, and RS-250 Reaction Station Accessories