missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Monoclonal Antibody (INS05 (2D11-H5)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512037
This item is not returnable.
View return policy
Description
MA5-12037 targets Insulin in IHC (P) applications and shows reactivity with Human, Porcine, and Rat samples. The MA5-12037 immunogen is purified Insulin protein.
Insulin Monoclonal antibody specifically detects Insulin in Human, Porcine, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
Insulin | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P01308, P01315, P01322, P01323 | |
INS, Ins1, Ins2 | |
Purified Insulin protein | |
500 μL | |
Primary | |
Human, Porcine, Rat | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
INS05 (2D11-H5) | |
Unconjugated | |
INS | |
AA986540; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2; Ins-2; Ins2-rs1; InsII; Insulin; insulin 1; insulin 2; Insulin A chain; Insulin B chain; insulin I; insulin II; insulin-1; Insulin-1 A chain; Insulin-1 B chain; Insulin-2; Insulin-2 A chain; Insulin-2 B chain; IRDN; Mody; MODY10; Mody4; preproinsulin; proinsulin | |
Mouse | |
Protein G | |
RUO | |
24505, 24506, 3630, 397415 | |
4° C | |
Liquid |