Learn More
General Tools Round Glass Inspection Mirror
View and retrieve objects in hard-to-reach areas with this Round Glass Inspection Mirror.
Brand:  General Tools 09914-00
Product Code. 11729849
  • 233.00 NOK / Each
Only null left
Estimated Shipment: 08-09-2025
Description
  • Telescopic shafts allow extended reach.
  • Adjustable mirror.
  • These inspection tools offer telescopic shafts that allow extended reach to inaccessible spaces.
  • Mirrors adjust to any angle for visual examination in trouble areas.
  • Magnets, mirrors 09914-00 and 09914-04, and inspection kit feature a heavy-duty pocket clip.
Specifications
Specifications
1 Ea.
31.75 mm
Used for visual examination in trouble areas
Round
Glass
1.25 in.
Inspection Mirror
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
General Tools Round Glass Inspection Mirror > Round Glass Inspection Mirror; Dia.: 31.75 mm; Length: 139.7 to 685.8 mm; Shape: Round