missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Insert

Insert E 118 for the optimum loading of half petri dishes

Brand:  Miele Professional™ 3830270

Product Code. 10407872

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Suitable for 38 half petri dishes with Ø 100 mm
  • equipped with 38 holders, height 70 mm, distance approx. 26 mm
  • For use in upper basket
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stainless Steel
Stainless Steel
Washer-disinfectors, laboratory
120 mm
Insert/Module for Petri Dishes
38 petri dishes
445 mm
460 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Miele Professional™ Insert >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.