Learn More
Kartell™ Inoculation Loops and Needles
Manufactured with high impact polystyrene for good dimensional stability. Kartell™ Inoculation Loops and Needles provide a two–in–one sterile disposable tool for inoculation.
Brand:  Kartell™ 0265000
Product Code. 10789651
  • 6335.00 NOK / Pack of 8000
Only null left
Estimated Shipment: 26-06-2025
Description
  • Gamma sterilized and packed in polybags
  • Looped end to retrieve an inoculum from a culture, transfer inoculum for streaking or transfer microscopic organisms
  • Needle end for picking off individual isolated colonies  or making stab inoculations into agar slants or tubes of solid culture medium
  • CE–IVD
Specifications
Specifications
Yes
Sterile
Neutral
Polybags
High impact polystyrene
1μL
2000 Pk.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kartell™ Inoculation Loops and Needles > Capacity: 1μL