Learn More
3M™ Inner Cover Plate
Brand:  3M™ 7000000182
185.70 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11765284
  • 264.00 NOK / Pack of 5
Estimated Shipment: 07-07-2025
to see stock.
Explore more special offers
Specifications
Inner Cover Plate
42 x 91 mm
Product Title
3M™ Inner Cover Plate > Size: 42 x 91mm