Learn More
B Braun™ Injekt Solo Cone Syringes
Enjoy the smooth, steady operation of the B Braun™ Solo Cone Leur syringe. Its green plunger enables easy recognition of filling volumes and its double-seal stopper keeps contents intact.
Brand:  B Braun™ 4606027V
 View more versions of this product
Product Code. 12722637
  • 82.00 NOK / Pack of 100
Edge
Description
Description
Enjoy the smooth, steady operation of the B Braun™ Solo Cone Leur syringe. Its green plunger enables easy recognition of filling volumes and its double-seal stopper keeps contents intact.
  • Injekt™
  • Transparent barrel with graduations
  • Green plunger
  • Barrel and plunger rod: Polypropylene
  • No DEHP or natural rubber
Specifications
Specifications
EN ISO 7886-1
Single-use
2 mL
Syringe
1/Ea.
Sterile
Certificates
Promotions
Promotions
Product Content Correction
Product Title
B Braun™ Injekt Solo Cone Syringes > Capacity: 2mL, Sterile