Learn More
3M™ Inhalation Valve Product Code.: 15729657
Estimated Shipment: 28-08-2025
Only null left
Add to basket
3M™ Inhalation Valve
Product Code. 15729657
Color: Light Blue
Click to view available options
:
Color: Light Blue
Product Code. 15729657
Brand: 3M™ 7000029731
Only null left
Estimated Shipment: 28-08-2025
Add to basket
Only null left
Add to basket
3M™ Inhalation Valve is suitable for use with 3M™ Reusable Half Mask 7500 Series.
  • Replacement inhalation valve for half mask respirator
  • Replace damaged parts to ensure correct levels of protection and comfort
  • For use with 3M™ Half Face Masks 7500 Series
TRUSTED_SUSTAINABILITY
Specifications
Color Light Blue
Product Type Inhalation Valve
For Use With (Application) For all painting work, mixing of paints and use of adhesives
For Use With (Equipment) Half Mask Series 7500
Product Content Correction
Product Title
3M™ Inhalation Valve >