missing translation for 'onlineSavingsMsg'
Learn More

Influenza A H5N1 Hemagglutinin Rabbit anti-Influenza virus, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

2780.00 NOK - 4640.00 NOK

Specifications

Antigen Influenza A H5N1 Hemagglutinin
Dilution Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18337892
View Documents
Bio-Techne
NBP3-12806-50UL
50 μg
2780.00 NOK
50µL
Please to purchase this item. Need a web account? Register with us today!
18322712
View Documents
Bio-Techne
NBP3-12806-100UL
100 μg
4640.00 NOK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Influenza A H5N1 Hemagglutinin Polyclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Influenza virus samples. It is validated for Western Blot, ELISA
Specifications

Specifications

Influenza A H5N1 Hemagglutinin
Western Blot, ELISA
Unconjugated
Rabbit
Immunology, Virology Bacteria and Parasites
0.2μm filtered solution in PBS without Preservative
IgG
Protein A purified
Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
Influenza virus
Purified, recombinant Influenza A H5N1 Hemagglutinin (ACI06178.1; Met1-Gln531)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.