missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H3N2 Hemagglutinin Rabbit anti-Influenza virus, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
2780.00 NOK - 4640.00 NOK
Specifications
Antigen | Influenza A H3N2 Hemagglutinin |
---|---|
Dilution | Western Blot 1:1000-1:5000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18329624
|
Bio-Techne
NBP3-12805-50UL |
50 μg |
2780.00 NOK
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18395494
|
Bio-Techne
NBP3-12805-100UL |
100 μg |
4640.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Influenza A H3N2 Hemagglutinin Polyclonal antibody specifically detects Influenza A H3N2 Hemagglutinin in Influenza virus samples. It is validated for Western BlotSpecifications
Influenza A H3N2 Hemagglutinin | |
Western Blot | |
Unconjugated | |
Rabbit | |
Virology Bacteria and Parasites | |
0.2μm filtered solution in PBS without Preservative | |
IgG | |
Protein A purified |
Western Blot 1:1000-1:5000 | |
Polyclonal | |
Purified | |
RUO | |
Influenza virus | |
Purified, recombinant Influenza A H3N2 Hemagglutinin (AAA43178.1; Met1-Arg345) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title