missing translation for 'onlineSavingsMsg'
Learn More

Indole-5-carboxylic acid, 98%

CAS: 1670-81-1 | C9H6NO2 | 160.15 g/mol

514.00 NOK - 1311.74 NOK

Chemical Identifiers

CAS 1670-81-1
Molecular Formula C9H6NO2
Molecular Weight (g/mol) 160.15
MDL Number MFCD00005678
InChI Key IENZCGNHSIMFJE-UHFFFAOYSA-M
Synonym indole-5-carboxylic acid, 5-carboxyindole, 5-indolecarboxylic acid, indole-5-carboxylic aicd, pubchem1694, 5-carboxy-1h-indole, indole-5-carboxylicacid, indol-5-carboxylic acid, 5-indole carboxylic acid, indole 5-carboxylic acid
PubChem CID 74280
IUPAC Name 1H-indole-5-carboxylic acid
SMILES [O-]C(=O)C1=CC=C2NC=CC2=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10489610
SDS View Documents
Thermo Scientific Acros
122210010
1 g Glass bottle
514.00 NOK
1g
Please to purchase this item. Need a web account? Register with us today!
10696312
SDS View Documents
Thermo Scientific Acros
122210050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indole-5-carboxylic acid, 98%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.