missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indane, 95%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -51°C |
Color | Colorless to Yellow |
Density | 0.9600g/mL |
Boiling Point | 176°C |
Flash Point | 50°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C9H10 |
Refractive Index | 1.5350 to 1.538 |
Show More |
Safety and Handling

Product Identifier
- Indan
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Indane, 95% >
Spot an opportunity for improvement?Share a Content Correction