missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert Incubator Im IF55m, 53L, +20 to +80 °C
Brand: Memmert IF55M
Incubator Im IF55m, 53L, +20 to +80 °C
This item is not returnable.
View return policy
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert Incubator Im IF55m, 53L, +20 to +80 °C >
Spot an opportunity for improvement?Share a Content Correction