Learn More
IMPA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16593-1-AP-20UL
 View more versions of this product
Product Code. 16856254
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
IMPA1, also known as Myo-inositol monophosphatase 1, is responsible for the procurement of inositol that is required for synthesis of phosphatidylinositol and polyphosphoinositides. IMPA1 exists as a homodimer and has been identified as the pharmacological target for lithium action in the brain. IMPA1 is the principal enzyme of the phosphatidyl inositol signaling pathway, and inhibition of inositol monophosphatase hydrolysis may underlie the anti-manic and anti-depressant actions of Li+. Studies indicate that a variation in the 277 codon coding region of the IMPA1 gene has not been observed in manic-depressive patients, therefore suggesting that polymorphisms or mutations in the noncoding regions of this gene may influence the lithium response in psychiatric patients.
Specifications
Specifications
IMPA1
Polyclonal
Unconjugated
IMPA1
IMP 1, IMPA, IMPA1, IMPase 1, Inositol monophosphatase 1
Rabbit
Antigen Affinity Chromatography
RUO
3612, 55980, 83523
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O55023, P29218, P97697
IMPA1
IMPA1 Fusion Protein Ag9632
20 μL
Primary
Human, Rat, Mouse