Learn More
STEMCELL Technologies ImmunoCult™ Human CD3/CD28/CD2 T Cell Activator

Human T cell activation and expansion reagent
3451.49 NOK - 16584.86 NOK
Specifications
Cell Type | T Cells, T Cells, CD4+, T Cells, CD8+ |
---|---|
For Use With (Application) | Activation, Cell Culture, Expansion |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15585273
![]() |
STEMCELL Technologies
10970 |
2 mL |
3451.49 NOK
2mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15531835
![]() |
STEMCELL Technologies
10990 |
5 x 2 mL |
16584.86 NOK
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Achieve robust activation and expansion of T cells in the absence of magnetic beads, feeder cells, or antigens
- This product’s gentle activation stimulus ensures a high viability of activated T cells, which can be further expanded in ImmunoCult™-XF T Cell Expansion Medium (Catalog #10981) or other media for culturing human T cells
- ImmunoCult™ Human CD3/CD28/CD2 T Cell Activator consists of soluble tetrameric antibody complexes that bind to and cross-link CD3, CD28, and CD2 cell surface ligands, thereby providing the required primary and co-stimulatory signals for T cell activation
- This product is designed for cell therapy research applications, but may be qualified for use as an ancillary material (AM) following the framework outlined in USP<1043>
- Contact STEMCELL Technologies to learn how we can work with you to qualify this reagent as an AM under an approved investigational new drug (IND) or clinical trial application (CTA)
Specifications
T Cells, T Cells, CD4+, T Cells, CD8+ | |
Activation, Cell Culture, Expansion |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.