missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ ImmunO™ Prostate Specific Antigen (PSA)

Filter sterilized

Brand:  MP Biomedicals™ 08322601

Product Code. 11443596

  • 3392.00 NOK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Sourced from human seminal fluid.

  • Purity: >96% by SDS-PAGE
  • Activity: 0.5-5mg PSA/ml
  • pH:7.5
  • Concentration:1mg/ml
  • pI: 6.2-6.5
  • Supplied as solution in 0.05M sodium phosphate buffer containing 0.15M NaCl and 0.09% sodium azide
  • Found negative for HIV 1 and 2 antibodies, Hepatitis B surface antigen and HCV antibodies
  • Sodium azide is added as preservative
  • This product is a biohazard

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1mg/mL
ImmunO™ Prostate Specific Antigen
100μg
0° to 5°C
Cell Biology and Analysis
7.5
Human Seminal Fluid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers