Learn More
Cytiva Immobiline™ DryStrip Kit and Components
Precast Immobiline DryStrip gels for running the first dimension of a 2-D electrophoresis experiment with the Multiphor II electrophoresis system
1465.00 NOK - 36805.00 NOK
Specifications
For Use With (Equipment) Multiphor II Electrophoresis System
View More Specs
Products 7
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
10310695
View accessory products View Documents
Cytiva
18-1004-30
Immobiline DryStrip Kit
36805.00 NOK
1 set
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
10565585
View Documents
Cytiva
18-1004-31
Tray and Electrode Holder
24015.00 NOK
Each
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
11540694
View Documents
Cytiva
18-1004-35
Sample Cup
1465.00 NOK
Pack of 60
In Stock
to see stock.
Add to basket
10176674
View Documents
Cytiva
18-1018-66
Anode
7190.00 NOK
1 set
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
10489404
View Documents
Cytiva
18-1004-33
Sample Cup Bar
2260.00 NOK
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
10239624
View Documents
Cytiva
18-1004-34
Dry Strip Slinger
1780.00 NOK
Pack of 4
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
10391315
View Documents
Cytiva
18-1018-67
Cathode
7190.00 NOK
1 set
Estimated Shipment: 13-02-2025
Description
Run up to 12 Immobiline DryStrip strips simultaneously with up to 100μL on each Immobiline DryStrip.
  • The instruction manual presents all necessary information for the entire method for horizontal 2-D electrophoresis. By appropriate adaptations, the SDS electrophoresis can also be performed using vertical electrophoresis equipment
  • Immobiline DryStrip kit facilitates sample application and running and equilibration of Immobiline DryStrip for the first dimension in 2-D (two-dimensional) electrophoresis
  • Isoelectric focusing with an immobilized pH gradient (IPG), makes true isoelectric focusing possible and significantly improves the reproducibility of the spot distribution along the pH gradient axis of 2-D maps
  • Individual components are also available separately
Specifications
Specifications
Multiphor II Electrophoresis System