Learn More
Imidazo[1,2-a]pyridine-7-carbohydrazide, 95% Product Code.: 15447507
Imidazo[1,2-a]pyridine-7-carbohydrazide, 95%
Product Code. 15447507
1 g, 1g
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15447507
Brand: Thermo Scientific Alfa Aesar H35365.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 421595-78-0 | C8H8N4O | 176.179 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 421595-78-0
Molecular Formula C8H8N4O
Molecular Weight (g/mol) 176.179
MDL Number MFCD13183226
InChI Key VXJBLZXLLRYHPD-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-7-carbohydrazide, imidazo 1,2-a pyridine-7-carboxylic acid hydrazide, 7-hydrazinocarbonyl imidazo 1,2-a pyridine
PubChem CID 55281632
IUPAC Name imidazo[1,2-a]pyridine-7-carbohydrazide
SMILES C1=CN2C=CN=C2C=C1C(=O)NN
Specifications
Melting Point 258°C to 261°C
Quantity 1 g
Formula Weight 176.18
Percent Purity 95%
Chemical Name or Material Imidazo[1,2-a]pyridine-7-carbohydrazide
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Product Code
Lot Number10179514 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH35365.MD,H35365.03
Product Title
Imidazo[1,2-a]pyridine-7-carbohydrazide, 95% >