missing translation for 'onlineSavingsMsg'
Learn More

IL-9 Mouse anti-Human, CoraLite 594, Clone: 1F3A11, Proteintech

Mouse Monoclonal Antibody

1565.79 NOK - 4225.34 NOK

Specifications

Antigen IL-9
Clone 1F3A11
Applications Immunofluorescence
Classification Monoclonal
Conjugate CoraLite 594
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17711419
View Documents Promotion Details
Proteintech
CL594-66144-20UL
20 μL
1565.79 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
17669956
View Documents Promotion Details
Proteintech
CL594-66144-100UL
100 μL
4225.34 NOK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-9 Monoclonal antibody apecifically detetects IL-9 antigen in Human Samples. It is validated for Immunofluorescence.
Specifications

Specifications

IL-9
Immunofluorescence
CoraLite 594
Mouse
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
3578
IL-9 fusion protein Ag21435
Primary
Store at -20°C. Avoid exposure to light.
1F3A11
Monoclonal
Liquid
Human
BC066284
IL9
IgG2b
Protein A purification
Videos
SDS
Documents

Documents

Product Certifications