Learn More
IL-23R Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27163-1-AP-150UL
This item is not returnable.
View return policy
Description
The IL-23R antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human IL-23R. This antibody recognizes human, mouse antigen. The IL-23R antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
IL-23R | |
Polyclonal | |
Unconjugated | |
150 μL | |
149233 | |
IgG |
Western Blot, Immunohistochemistry | |
300 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |